Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IKK epsilon Rabbit anti-Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548682

Catalog No. PA548682

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

IKBKE is a noncanonical I-kappa-B kinase that is essential for regulating antiviral signaling pathways. IKBKE has also been identified as a breast cancer oncogene and is amplified and overexpressed in over 30% of breast carcinomas and breast cancer cell lines .
Specifications

Specifications

IKK epsilon
Polyclonal
PBS with 0.09% sodium azide
Q9R0T8
Inhibitor of nuclear factor kappa-B kinase subunit epsilon, I-kappa-B kinase epsilon, IKK-E, IKK-epsilon, IkBKE, Inducible I kappa-B kinase, IKK-i, Ikbke, Ikke, Ikki
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
56489
Western Blot
Unconjugated
Ikbke
Liquid
IKBKE
KLH conjugated synthetic peptide between 470-498 amino acids from the Central region of mouse Ikbke.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit