Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IKK alpha Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA587434
Description
Positive Samples: Jurkat, HeLa, HepG2; Cellular Location: Cytoplasm, Nucleus
NFkB comprises a family of cellular transcription factors that are involved in the inducible expression of a variety of cellular genes that regulate the inflammatory response and control of cell death. In the cytoplasm NFk B is negatively modulated by the inhibitory proteins Ik-B. In turn Ik-B is phosphorylated by a cellular kinase complex called IKK. IKK is a heterodimer composed of two kinases: IKK alpha and IKK beta which phosphorylate Ik-B leading to its degradation and the resulting translocation of NFk B to the nucleus. IKK kinase activity is modulated negatively by pharmaceutical agents such as aspirin and positively by various cellular components such as TNF alpha , endotoxins and overexpression of cellular kinases like MEKK1. Aspirin appears to have its effect by inhibiting the binding of ATP to IKK.Specifications
IKK alpha | |
Polyclonal | |
Unconjugated | |
CHUK | |
CHUK | |
Recombit protein of human CHUK | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunofluorescence, Western Blot | |
7.51 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O15111, Q60680 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1147, 12675 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title