Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IKK epsilon/IKBKE Mouse anti-Human, Alexa Fluor™ 647, Clone: OTI3C5, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271490AF647

Catalog No. N271490A647


Add to cart

Description

Description

IKK epsilon/IKBKE Monoclonal antibody specifically detects IKK epsilon/IKBKE in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

IKK epsilon/IKBKE
OTI3C5
Western Blot, Immunohistochemistry
Affinity Purified, Conjugated
IKBKE
Recombinant protein fragment corresponding to amino acids 474-716 of human IKK epsilon/IKBKE (NP_054721) produced in E.coli.
Immunogen affinity purified
Cancer, Phospho Specific, Protein Kinase
Primary
9641.0
Immunohistochemistry, Western Blot
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
EC 2.7.11, EC 2.7.11.10, I-kappa-B kinase epsilon, IkBKE, IKK-E, IKKEMGC125297, IKK-epsilon, IKKI, IKK-iMGC125295, Inducible I kappa-B kinase, inducible IkappaB kinase, inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase epsilon, inhibitor of nuclear factor kappa-B kinase subunit epsilon, KIAA0151IKK-related kinase epsilon, MGC125294
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit