Learn More
IL-1 beta/IL-1F2 Mouse anti-Human, Alexa Fluor™ 532, Clone: 43N1F9, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP227340AF532
Catalog No. NP227340A32
Add to cart
Description
Description
IL-1 beta/IL-1F2 Monoclonal antibody specifically detects IL-1 beta/IL-1F2 in Human samples. It is validated for Western Blot.
Specifications
Specifications
IL-1 beta/IL-1F2
43N1F9
Western Blot
catabolin, IL-1, IL-1 beta, IL-1B, IL1-BETA, IL1F2IL-1 beta, interleukin 1, beta, interleukin-1 beta, preinterleukin 1 beta, pro-interleukin-1-beta
Mouse
IgG1 Kappa
Store at 4°C in the dark.
Monoclonal
Human
Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
IL1B
Full-length recombinant human IL-1 beta protein was used as immunogen for this antibody.
0.1 mL
Primary
3553