Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-1 beta/IL-1F2 Mouse anti-Human, Rat, Alexa Fluor 532, Clone: 43N1G6, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP227342AF532
Description
IL-1 beta/IL-1F2 Monoclonal specifically detects IL-1 beta/IL-1F2 in Human, Rat samples. It is validated for Western Blot.Specifications
IL-1 beta/IL-1F2 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
IL1B | |
Full-length recombinant human IL-1beta protein was used as immunogen for this antibody. | |
0.1 mL | |
Primary | |
Human, Rat | |
IgG1 κ |
Western Blot | |
43N1G6 | |
Western Blot | |
catabolin, IL-1, IL-1 beta, IL-1B, IL1-BETA, IL1F2IL-1 beta, interleukin 1, beta, interleukin-1 beta, preinterleukin 1 beta, pro-interleukin-1-beta | |
Mouse | |
Protein G purified | |
Adaptive Immunity, Apoptosis, Biologically Active Proteins, Cancer, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Innate Immunity, Metastasis | |
3553.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title