Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-1 RI Rabbit anti-Human, Clone: 24, Novus Biologicals™
Rabbit Monoclonal Antibody
$321.00 - $493.00
Specifications
Antigen | IL-1 RI |
---|---|
Clone | 24 |
Dilution | ELISA 1:5000-1:10000 |
Applications | ELISA |
Classification | Monoclonal |
Description
IL-1 RI Monoclonal specifically detects IL-1 RI in Human samples. It is validated for ELISA.Specifications
IL-1 RI | |
ELISA 1:5000-1:10000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CD121 antigen-like family member A, CD121a antigen, IL1R, IL1R1, IL1RT1, IL-1RT1, IL-1RT-1, Interleukin 1 Receptor 1, interleukin 1 receptor, type I, interleukin receptor 1, interleukin-1 receptor type 1, p80 | |
Recombinant Human IL-1 RI Protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
24 | |
ELISA | |
Unconjugated | |
Rabbit | |
Cancer, Cytokine Research, Immunology, Innate Immunity, Phospho Specific | |
0.2 um filtered solution in PBS | |
3554 | |
IgG | |
Protein A purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title