Learn More
IL-1 RI Rat anti-Mouse, Alexa Fluor 700, Clone: RM0013-11B7, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NB11085471AF700
Catalog No. NBT85471700
Add to cart
Description
Description
IL-1 RI Monoclonal antibody specifically detects IL-1 RI in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
IL-1 RI
RM0013-11B7
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
IL1R1
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat) immunized with mouse recombinant extracellular domain of mouse IL1 Receptor 1 Antibody (RM0013-11B7).
Protein G purified
Cancer, Cytokine Research, Immunology, Innate Immunity, Phospho Specific
Primary
3554
Mouse
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
CD121 antigen-like family member A, CD121a antigen, IL1R, IL1R1, IL1RT1, IL-1RT1, IL-1RT-1, Interleukin 1 Receptor 1, interleukin 1 receptor, type I, interleukin receptor 1, interleukin-1 receptor type 1, p80
Rat
IgG2
0.1 ml
Store at 4C in the dark.
Monoclonal
This is recognizes the extracellular domain of mouse IL1 Receptor 1 Antibody (RM0013-11B7)