Learn More
IL-1 RI Rat anti-Mouse, Alexa Fluor 750, Clone: 129304, R&D Systems™
Rat MonoclonalAntibody
Manufacturer:  R&D Systems FAB7712S100UG
Catalog No. FAB7712S100
Add to cart
This item is not returnable. View return policy
Description
Description
IL-1 RI Monoclonal antibody specifically detects IL-1 RI in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
IL-1 RI
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
P13504
IL1R1
Mouse myeloma cell line NS0-derived recombinant mouse IL-1 RI Leu20-Lys338 Accession # P13504
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects mouse IL-1 R1 in direct ELISAs and Western blots.
Flow Cytometry
129304
Alexa Fluor 750
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
CD121 antigen-like family member A, CD121a, CD121a antigen, IL1R, IL1R1, IL1RI, IL-1RI, IL1RT1, IL-1RT1, IL-1RT-1, Interleukin 1 Receptor 1, interleukin 1 receptor, type I, interleukin receptor 1, interleukin-1 receptor type 1, p80
Rat
IgG2b
RUO
Primary
3554
Mouse
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)