Learn More
IL-1 RII Mouse anti-Human, DyLight 550, Clone: MM0356-4T28, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP211646R
Catalog No. NBP211646R
Add to cart
Description
Description
IL-1 RII Monoclonal antibody specifically detects IL-1 RII in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
Specifications
IL-1 RII
MM0356-4T28
Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen
Purified
IL1R2
Produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with human recombinant protein of IL-1 R II extracellular domain.
Protein G purified
Cancer, Phospho Specific, Signal Transduction
Primary
7850
Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
DyLight 550
50mM Sodium Borate with 0.05% Sodium Azide
beta, CD121b, CD121b antigen, IL1RBCD121 antigen-like family member B, IL-1R-beta, IL-1RT2, IL-1RT-2, interleukin 1 receptor, type II, Interleukin-1 receptor beta, MGC47725
Mouse
IgG2
0.1 ml
Store at 4C in the dark.
Monoclonal
Human