Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-1 RII Rabbit anti-Rat, Alexa Fluor 700, Clone: 21, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290886AF700
Description
IL-1 RII Monoclonal specifically detects IL-1 RII in Rat samples. It is validated for ELISA.Specifications
IL-1 RII | |
Monoclonal | |
Alexa Fluor 700 | |
beta, CD121b, CD121b antigen, IL1RBCD121 antigen-like family member B, IL-1R-beta, IL-1RT2, IL-1RT-2, interleukin 1 receptor, type II, Interleukin-1 receptor beta, MGC47725 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Rat IL-1 RII (P43303; Met1-Glu355). | |
0.1 mL | |
Cancer, Phospho Specific, Signal Transduction | |
7850 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
21 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Rat | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title