Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-1 RII Rabbit anti-Rat, Clone: 21, Novus Biologicals™

Rabbit Monoclonal Antibody
$342.00 - $524.00
Specifications
Antigen | IL-1 RII |
---|---|
Clone | 21 |
Dilution | ELISA 1:5000-1:10000 |
Applications | ELISA |
Classification | Monoclonal |
Description
IL-1 RII Monoclonal specifically detects IL-1 RII in Rat samples. It is validated for ELISA.Specifications
IL-1 RII | |
ELISA 1:5000-1:10000 | |
Monoclonal | |
Purified | |
RUO | |
Rat | |
beta, CD121b, CD121b antigen, IL1RBCD121 antigen-like family member B, IL-1R-beta, IL-1RT2, IL-1RT-2, interleukin 1 receptor, type II, Interleukin-1 receptor beta, MGC47725 | |
Recombinant Rat IL-1 RII Protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
21 | |
ELISA | |
Unconjugated | |
Rabbit | |
Cancer, Phospho Specific, Signal Transduction | |
0.2 um filtered solution in PBS | |
7850 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title