Learn More
IL-1 RII Rat anti-Mouse, DyLight 680, Clone: RM0014-7D7, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NB11085472FR
Catalog No. NBT85472FR
Add to cart
Description
Description
IL-1 RII Monoclonal antibody specifically detects IL-1 RII in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
IL-1 RII
RM0014-7D7
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
IL1R2
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse Recombinant extracellular domain of mouse IL1R-II protein.
Protein G purified
Cancer, Phospho Specific, Signal Transduction
Primary
7850
Mouse
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
DyLight 680
50mM Sodium Borate with 0.05% Sodium Azide
beta, CD121b, CD121b antigen, IL1RBCD121 antigen-like family member B, IL-1R-beta, IL-1RT2, IL-1RT-2, interleukin 1 receptor, type II, Interleukin-1 receptor beta, MGC47725
Rat
IgG2
0.1 ml
Store at 4C in the dark.
Monoclonal
This recognizes extracellular domain of mouse IL1R-II protein.