Learn More
Novus Biologicals™ IL-10 ELISA Kit [Biotin]
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 
Manufacturer:  Novus Biologicals™ NBP192676
Catalog No. NBP192676
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
The mouse IL-10 ELISA is an enzyme-linked immunosorbent assay for the quantitative detection of mouse IL-10
Specifications
Specifications
15.6 - 1,000pg/mL
3586
ELISA Kit
5.0pg/mL
Storage of components varies. See protocol for specific instructions.
Adaptive Immunity, Apoptosis, Asthma, Cancer, Cytokine Research, Immunology, Innate Immunity
CSIFMGC126450, Cytokine synthesis inhibitory factor, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF
ELISA
Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to mouse IL-10, Biotin-Conjugate anti-mouse IL-10 polyclonal antibody, Streptavidin-HRP, Mouse IL-10 Standard lyophilized, 2ng/mL upon reconstitution, Sample Diluent, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films
96 Tests
Cell culture supernatant, serum, plasma (EDTA, citrate)
IL-10
IL10