Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-10 Mouse anti-Human, Unconjugated, Clone: 2H6, MilliporeSigma™

Mouse Monoclonal Antibody

$147.00 - $372.00

Specifications

Antigen IL-10
Applications Immunohistochemistry (Paraffin)
Conjugate Unconjugated
Format Purified
Host Species Mouse
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABF22931MI
View Documents
MilliporeSigma
MABF2293100UG
100 μg
Each for $372.00
Add to cart
 
MABF22932MI
View Documents
MilliporeSigma
MABF229325UG
25 μg
Each for $147.00
Add to cart
 
Description

Description

IL-10 Monoclonal antibody specifically detects IL-10 in Human samples. It is validated for Immunohistochemistry (Paraffin), Affinity Binding Assay.
Specifications

Specifications

IL-10
Unconjugated
Mouse
Interleukin-10;Cytokine synthesis inhibitory factor;CSIF
Full-length recombinant mature human IL-10 (without the signal peptide).
Primary
Stable for 1 year at 2 to 8°C from date of receipt.
Immunohistochemistry (Paraffin)
Purified
Human
IL10
Monoclonal
Protein G Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit