Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-10 Mouse anti-Rat, Bovine, Equine, Goat, Sheep, Clone: CC320, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 4 publications
$176.85 - $493.00
Specifications
Antigen | IL-10 |
---|---|
Clone | CC320 |
Concentration | 1 mg/mL |
Applications | Flow Cytometry, ELISA, Functional Assay, CyTOF |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10063026
|
Novus Biologicals
NB10063026 |
0.25 mg |
Each for $493.00
|
|
NB017532
|
Novus Biologicals
NB100630260.05MG |
0.05 mg |
Each for $176.85
|
|
Description
IL-10 Monoclonal specifically detects IL-10 in Rat, Bovine, Equine, Goat, Sheep samples. It is validated for Flow Cytometry, ELISA, Functional, CyTOF-ready.Specifications
IL-10 | |
1 mg/mL | |
Monoclonal | |
Purified | |
RUO | |
Bovine, Equine, Goat, Sheep | |
3586 | |
Plasmid cDNA encoding bovine IL-10 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
CC320 | |
Flow Cytometry, ELISA, Functional Assay, CyTOF | |
Unconjugated | |
Mouse | |
Adaptive Immunity, Apoptosis, Asthma, Cancer, Immunology, Innate Immunity | |
CSIFMGC126450, Cytokine synthesis inhibitory factor, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF | |
IL10 | |
IgG1 | |
Protein G purified | |
Clone CC320 has been shown to neutralise the activity of bovine IL-10 as measured by the inhibition of the inhibitory activity of IL-10 on IFN gamma synthesis (2). |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title