Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-10R alpha Hamster anti-Mouse, FITC, Clone: RM0101-9E12, Novus Biologicals™
Hamster Monoclonal Antibody
Supplier: Novus Biologicals NBP122459F
Description
IL-10R alpha Monoclonal antibody specifically detects IL-10R alpha in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin).Specifications
IL-10R alpha | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
IL10RA | |
Mouse recombinant protein of the Extracellular domain of IL-10R alpha | |
0.1 mL | |
Primary | |
This was produced from a hybridoma (mouse myeloma fused with spleen cells from a hamster immunized with mouse recombinant protein of extracellular domain of IL-10R alpha. | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
RM0101-9E12 | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
CD210 antigen, CDw210a, HIL-10R, IBD28, IL-10 receptor subunit alpha, IL-10R subunit 1, IL-10R1, IL-10RA, IL10RIL-10R subunit alpha, interleukin 10 receptor, alpha, interleukin-10 receptor alpha chain, Interleukin-10 receptor subunit 1, interleukin-10 receptor subunit alpha | |
Hamster | |
Protein A or G purified | |
Cytokine Research | |
3587 | |
Mouse | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title