Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ IL-10 R beta Antibody Pair [HRP]

Polyclonal/Monoclonal Antibody Pair

Manufacturer:  Novus Biologicals™ NBP2793275PLATES

 View more versions of this product

Catalog No. NB009694


Add to cart

Description

Description

This solid phase sandwich ELISA utilizes an antibody specific for Human IL-10 R beta.
Specifications

Specifications

Sandwich ELISA
Antibody Pairs
78.125 pg/mL
3588
RUO
IL-10 R beta
Cytokine Research, Immunology, Innate Immunity
Human
IL10RB
1. Capture Antibody: Dilute to a working concentration of 2.0 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to a working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute standard powder with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve usi ng 2-fold serial dilutions in sample dilution buffer, and a high standard of 5000 pg/mL is recommended.
HRP
This solid phase sandwich ELISA utilizes an antibody specific for Human IL-10 R beta.
Sandwich ELISA
  • Detection Antibody: 0.7 mg/mL rabbit anti-IL-10 R beta polyclonal antibody conjugated to horseradish-peroxidase (HRP).
  • Capture Antibody: 0.5 mg/mL of mouse anti-IL-10 R beta monoclonal antibody.
  • Standard: Each vial contains 110 ng of recombinant IL-10 R beta.
  • Storage is content dependent.
    Rabbit/Mouse
    5 Plates
    78.13-5000 pg/mL
    CDw210b antigen, CRF2-4IL-10R subunit 2, CRFB4human transmembrane receptor protein; cytokine receptor10CDW210B, D21S58, D21S66, IBD25, IL-10 receptor subunit beta, IL-10R2IL-10R subunit beta, IL-10RB, interleukin 10 receptor, beta, Interleukin-10 receptor subunit 2, interleukin-10 receptor subunit beta, member 4
    SDS
    Documents
    Product Certifications
    Promotions

    Promotions

    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit