Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ IL-10 R beta Recombinant Protein

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

$287.00 - $1025.00

Specifications

Formulation PBS (pH 7.4) containing 10% glycerol. containingno Preservative
Gene Alias CDw210b antigen, CRF2-4IL-10R subunit 2, CRFB4human transmembrane receptor protein; cytokine receptor10CDW210B, D21S58, D21S66, IBD25, IL-10 receptor subunit beta, IL-10R2IL-10R subunit beta, IL-10RB, interleukin 10 receptor, beta, Interleukin-10 receptor
Gene ID (Entrez) 3588
Gene Symbol IL10RB
Name Recombinant IL-10 R beta Protein
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB60218250U View Documents Novus Biologicals™
NBP260218250UG
250μg Each for $1,025.00
Add to cart
 
NB6021850UG View Documents Novus Biologicals™
NBP26021850UG
50μg Each for $287.00
Add to cart
 
Description

Description

A recombinant protein with a C-Terminal hIgG-His-tag and corresponding to the amino acids 20-220 of Human IL-10 R beta The Recombinant Human IL-10 R beta Protein is derived from Baculovirus. The Recombinant Human IL-10 R beta Protein has been validated for the following applications: SDS-Page.
Specifications

Specifications

PBS (pH 7.4) containing 10% glycerol. containingno Preservative
3588
Recombinant IL-10 R beta Protein
Baculovirus
1mg/mL
SDS-Page
CDw210b antigen, CRF2-4IL-10R subunit 2, CRFB4human transmembrane receptor protein; cytokine receptor10CDW210B, D21S58, D21S66, IBD25, IL-10 receptor subunit beta, IL-10R2IL-10R subunit beta, IL-10RB, interleukin 10 receptor, beta, Interleukin-10 receptor
IL10RB
Recombinant Protein
RUO
< 1.0 EU per 1ug of protein (determined by LAL method)

For Research Use Only (RUO)

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit