Learn More
IL-10 Rat anti-Human,Virus, BB700, Clone: JES3-19F1, BD Horizon™
Rat Monoclonal Antibody
Manufacturer:  BD Biosciences 566568
Catalog No. BDB566568
Add to cart
Description
Description
IL-10 Monoclonal antibody specifically detects IL-10 in Human,Virus samples. It is validated for Flow Cytometry.
Specifications
Specifications
IL-10
JES3-19F1
Purified
IgG2a, κ
RUO
Primary
Human, Virus
Flow Cytometry
BB700
Rat
25 Tests
Refrigerate (2 to 8°C)
Monoclonal