Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-10R alpha Rat anti-Mouse, Clone: 1B1.3a, Novus Biologicals™

Rat Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen IL-10R alpha
Clone 1B1.3a
Concentration 0.2 mg/ml
Dilution Flow Cytometry 0.5-1 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml
Applications Flow Cytometry, Immunofluorescence
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122133
View Documents
Novus Biologicals
NBP30733010UG
100 μg
Each for $352.89
Add to cart
 
NB122134
View Documents
Novus Biologicals
NBP30733020UG
20 μg
Each for $138.37
Add to cart
 
Description

Description

IL-10R alpha Monoclonal antibody specifically detects IL-10R alpha in Mouse samples. It is validated for Flow Cytometry, Immunofluorescence
Specifications

Specifications

IL-10R alpha
0.2 mg/ml
Flow Cytometry, Immunofluorescence
Purified
RUO
10 mM PBS with 0.05% BSA
3587
IgG1 Kappa
Primary
Store at 4°C.
1B1.3a
Flow Cytometry 0.5-1 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml
Unconjugated
Rat
Mouse
CD210 antigen, CDw210a, HIL-10R, IBD28, IL-10 receptor subunit alpha, IL-10R subunit 1, IL-10R1, IL-10RA, IL10RIL-10R subunit alpha, interleukin 10 receptor, alpha, interleukin-10 receptor alpha chain, Interleukin-10 receptor subunit 1, interleukin-10 receptor subunit alpha
Purified recombinant extracellular region of Mouse IL-10R alpha
Monoclonal
Protein A or G purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit