Learn More
IL-10R alpha Armenian Hamster anti-Mouse, Janelia Fluor 549, Clone: RM0101-9E12, Novus Biologicals™
Hamster Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP122459JF549
Catalog No. NBP12459549
Add to cart
Description
Description
IL-10R alpha Monoclonal antibody specifically detects IL-10R alpha in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications
Specifications
IL-10R alpha
RM0101-9E12
Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
IL10RA
Mouse recombinant protein of the Extracellular domain of IL-10R alpha
Protein G purified
Store at 4°C in the dark.
Monoclonal
This was produced from a hybridoma (mouse myeloma fused with spleen cells from a hamster immunized with mouse recombinant protein of extracellular domain of IL-10R alpha.
Immunohistochemistry, Immunohistochemistry (Paraffin)
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
CD210 antigen, CDw210a, HIL-10R, IBD28, IL-10 receptor subunit alpha, IL-10R subunit 1, IL-10R1, IL-10RA, IL10RIL-10R subunit alpha, interleukin 10 receptor, alpha, interleukin-10 receptor alpha chain, Interleukin-10 receptor subunit 1, interleukin-10 receptor subunit alpha
Hamster
IgG2
0.1mL
Primary
3587
Mouse