Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-11R alpha Rabbit anti-Mouse, DyLight 350, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298219UV
Description
IL-11R alpha Polyclonal specifically detects IL-11R alpha in Mouse samples. It is validated for ELISA.Specifications
IL-11R alpha | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Mouse | |
Purified |
ELISA | |
DyLight 350 | |
AI314697, GP130, IL-11RA, Il11ra2, NR1 | |
Produced in rabbits immunized with purified, recombinant IL-11R alpha (NP_001156873.1; Met1-Gln367). | |
0.1 mL | |
Cytokine Research, Immunology, Innate Immunity | |
3590 | |
Store at 4°C in the dark. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title