Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-12/IL-23 p40 Rabbit anti-Human, Biotin, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299502B
Description
IL-12/IL-23 p40 Polyclonal specifically detects IL-12/IL-23 p40 in Human samples. It is validated for ELISA.Specifications
IL-12/IL-23 p40 | |
Polyclonal | |
PBS | |
Rabbit | |
Affinity purified | |
RUO | |
3593 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
Biotin | |
CLMF, CLMF p40, CLMF2, Cytotoxic lymphocyte maturation factor 40 kDa subunit, IL-12 subunit p40, IL-12BNK cell stimulatory factor chain 2, interleukin 12, p40, interleukin 12B (natural killer cell stimulatory factor 2, cytotoxic lymphocytematuration factor 2, p40), interleukin-12 beta chain, interleukin-12 subunit beta, natural killer cell stimulatory factor, 40 kD subunit, NKSF, NKSF2IL12, subunit p40 | |
Produced in rabbits immunized with purified, recombinant IL-12/IL-23 p40 (NP_002178.2). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title