Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-12 p40 Mouse anti-Human, Unconjugated, Clone: 11B10, MilliporeSigma™

Mouse Monoclonal Antibody

$147.00 - $372.00

Specifications

Antigen IL-12 p40
Applications Immunohistochemistry (Paraffin), Flow Cytometry
Conjugate Unconjugated
Format Purified
Host Species Mouse
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABF22951MI
View Documents
MilliporeSigma
MABF2295100UG
100 μg
Each for $372.00
Add to cart
 
MABF22952MI
View Documents
MilliporeSigma
MABF229525UG
25 μg
Each for $147.00
Add to cart
 
Description

Description

IL-12 p40 Monoclonal antibody specifically detects IL-12 p40 in Human samples. It is validated for Immunohistochemistry (Paraffin), Flow Cytometry.
Specifications

Specifications

IL-12 p40
Unconjugated
Mouse
Interleukin-12 subunit beta;IL-12B;Cytotoxic lymphocyte maturation factor 40 kDa subunit;CLMF p40;IL-12 subunit p40;NK cell stimulatory factor chain 2;NKSF2
His-tagged full-length human recombinant Interleukin-12 subunit beta (IL12 p40).
Primary
Stable for 1 year at 2 to 8°C from date of receipt.
Immunohistochemistry (Paraffin), Flow Cytometry
Purified
Human
IL12B, NKSF2
Monoclonal
Protein G Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit