Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-12 p70/IL-12A Mouse anti-Human, Alexa Fluor™ 594, Clone: 2Y37, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP222038AF594

Catalog No. NP222038A94


Add to cart

Description

Description

IL-12 p70/IL-12A Monoclonal antibody specifically detects IL-12 p70/IL-12A in Human samples. It is validated for Western Blot, Flow Cytometry.
Specifications

Specifications

IL-12 p70/IL-12A
2Y37
Western Blot, Flow Cytometry
Conjugated, Purified
IL12A
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with purified recombinant IL-12/IL-35 p35. Mouse anti human IL12A P29459.
Protein G purified
Alzheimers Research, Immunology, Innate Immunity, Neurodegeneration
Primary
3592.0
Flow Cytometry, Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
CLMF p35, Cytotoxic lymphocyte maturation factor 35 kDa subunit, IL-12 subunit p35, IL-12, subunit p35, interleukin 12, p35, interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocytematuration factor 1, p35), interleukin-12 alpha chain, interleukin-12 subunit alpha, natural killer cell stimulatory factor 1, 35 kD subunit, NF cell stimulatory factor chain 1, NK cell stimulatory factor chain 1, NKSF1, p35
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit