Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-12 p70 Mouse anti-Human, Clone: 2F7, MilliporeSigma™

Mouse Monoclonal Antibody

$145.80 - $373.00

Specifications

Antigen IL-12 p70
Clone 2F7
Concentration 1 mg/mL
Applications Flow Cytometry, Immunohistochemistry (Paraffin)
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABF2297100
View Documents
MilliporeSigma
MABF2297100UG
100 μg
Each for $373.00
Add to cart
 
MABF229725
View Documents
MilliporeSigma
MABF229725UG
25 μg
Each for $145.80
Add to cart
 
Description

Description

IL-12 p70 Monoclonal antibody specifically detects IL-12 p70 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin).
Specifications

Specifications

IL-12 p70
1 mg/mL
Unconjugated
Mouse
Human
His-tagged full-length recombinant human Interleukin-12A.
Monoclonal
Protein G purified
2F7
Flow Cytometry, Immunohistochemistry (Paraffin)
Purified
RUO
Purified mouse monoclonal antibody IgG1 in PBS with 0.05% sodium azide.
IgG1κ
Primary
Stable for 1 year at +2°C to +8°C from date of receipt.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit