Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-12 R beta 1 Mouse anti-Human, Alexa Fluor™ 647, Clone: OTI1A6, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271033AF647

Catalog No. N271033A647


Add to cart

Description

Description

IL-12 R beta 1 Monoclonal antibody specifically detects IL-12 R beta 1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

IL-12 R beta 1
OTI1A6
Western Blot
Affinity Purified, Conjugated
IL12RB1
Recombinant protein fragment corresponding to amino acids 33-283 of human IL-12 R beta 1 (NP_714912) produced in E.coli.
Immunogen affinity purified
Cell Cycle and Replication
Primary
3594.0
Western Blot
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
CD212, CD212 antigen, IL-12 receptor beta component, IL-12 receptor subunit beta-1, IL12R, IL-12R subunit beta-1, IL12RB, IL-12RB1, IL-12R-beta-1, IL-12R-BETA1, interleukin 12 receptor, beta 1, interleukin-12 receptor beta-1 chain, interleukin-12 receptor subunit beta-1, MGC34454
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit