Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-12 R beta 2 Antibody (305719), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB19592L
Description
IL-12 R beta 2 Monoclonal antibody specifically detects IL-12 R beta 2 in Human, Mouse samples. It is validated for Direct ELISASpecifications
IL-12 R beta 2 | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Direct ELISA | |
305719 | |
Direct ELISA | |
IL-12 receptor beta 2, IL-12 receptor subunit beta-2, IL-12R subunit beta-2, IL-12RB2, IL-12R-beta-2, interleukin 12 receptor, beta 2, interleukin-12 receptor beta-2 chain, interleukin-12 receptor subunit beta-2 | |
Mouse myeloma cell line NS0-derived recombinant human IL-12 R beta 2, Cys28-Asn622, Accession # Q99665 | |
0.1 mL | |
Cell Cycle and Replication | |
3595 | |
Store at 4C in the dark. | |
IgG1 |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction