Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-12 R beta 2 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP132558

Catalog No. NBP132558


Only null left
Add to Cart

Description

Description

IL-12 R beta 2 Polyclonal specifically detects IL-12 R beta 2 in Human samples. It is validated for Western Blot.
Specifications

Specifications

IL-12 R beta 2
Polyclonal
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
IL12RB2
Recombinant protein encompassing a sequence within the center region of human IL-12 Receptor beta2. The exact sequence is proprietary.
Affinity Purified
RUO
Primary
Tested in human. Not yet tested in any other species.
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Western Blot
1.0mg/mL
Western Blot 1:1000-1:10000
IL-12 receptor beta 2, IL-12 receptor subunit beta-2, IL-12R subunit beta-2, IL-12RB2, IL-12R-beta-2, interleukin 12 receptor, beta 2, interleukin-12 receptor beta-2 chain, interleukin-12 receptor subunit beta-2
Rabbit
97 kDa
100 μL
Cell Cycle and Replication
3595
Human
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only