Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-13 Mouse anti-Human, PE, Clone: 85BRD, eBioscience™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen 12713641

Catalog No. 50-112-2379


Add to cart

Description

Description

Description: The 85BRD monoclonal antibody reacts with human interleukin-13 (IL-13). IL-13, also known as NC30, is a 12.5 kDa protein secreted by activated T helper cells, CD8+ T cells, and NK cells. The biological activities of IL-13 include suppression of macrophage cytotoxic activity, upregulation of IL-1RA expression and suppression of inflammatory cytokine expression. Applications Reported: This 85BRD antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This 85BRD antibody has been pre-titrated and tested by intracellular staining and flow cytometric analysis of stimulated normal human peripheral blood cells using the Intracellular Fixation and Permeabilization Buffer Set (cat. 88-8824) and protocol. Please refer to Best Protocols section (Protocol A: Two step protocol for (cytoplasmic) intracellular proteins). This can be used at 5 µL (0.125 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 µm post-manufacturing filtered.

IL-13 is an immunoregulatory cytokine produced primarily by activated Th2 cells, and is involved in regulating inflammatory and immune responses. IL-13 down-regulates macrophage activity, thereby inhibiting the production of pro-inflammatory cytokines and chemokines, and is involved in allergen-induced asthma through mechanisms independent of IgE and eosinophils. IL-13 also regulates several stages of B cell maturation and differentiation, up-regulates CD23 and MHC class II expression, and promotes IgE isotype switching of B cells. Structurally, IL-13 closely resembles IL-3, IL-4, IL-5 and GM-CSF. The gene encoding IL-13, IL3, IL5, IL4, and CSF2 form a cytokine gene cluster on chromosome 5q, with the IL-13 gene particularly close to IL4.
Specifications

Specifications

IL-13
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P35225
Interleukin-13, ALRH antibody, NC30 antibody, P600 antibody
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
85BRD
PE
IL13
Liquid
IL13
IgG2b, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
3596
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.