Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ IL-13R alpha 1 Antibody Pair [HRP]

Mouse Monoclonal Antibody Pair

$293.00 - $629.00

Specifications

Target IL-13R alpha 1
Conjugate HRP
Target Species Human
Gene ID (Entrez) 3597
Gene Symbol IL13RA1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB009691
View Documents
Novus Biologicals™
NBP27932615PLATES
15 Plates
Each for $629.00
Add to cart
 
NB009692
View Documents
Novus Biologicals™
NBP2793265PLATES
5 Plates
Each for $293.00
Add to cart
 
Description

Description

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human IL-13R alpha 1.
Specifications

Specifications

IL-13R alpha 1
Human
IL13RA1
HRP
3597
Mouse
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit