Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-13R alpha 2 Rabbit anti-Rat, DyLight 350, Clone: 7, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290890UV
Description
IL-13R alpha 2 Monoclonal specifically detects IL-13R alpha 2 in Rat samples. It is validated for ELISA.Specifications
IL-13R alpha 2 | |
Monoclonal | |
DyLight 350 | |
cancer/testis antigen 19, CD213a2, CD213a2 antigen, CT19, IL-13 receptor subunit alpha-2, IL13BP, IL13R, IL-13R, IL-13R subunit alpha-2, IL-13RA2, IL-13R-alpha-2, interleukin 13 binding protein, interleukin 13 receptor alpha 2 chain, interleukin 13 receptor, alpha 2, interleukin-13 receptor subunit alpha-2, Interleukin-13-binding protein | |
This antibody was obtained from a rabbit immunized with purified, recombinant Rat IL-13R alpha 2 (Q8VHK6; Met1-Lys336). | |
0.1 mL | |
Cancer | |
3598 | |
Rat | |
Purified |
ELISA | |
7 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Has cross-reactivity in ELISA with: Mouse IL13RA2 No cross-reactivity in ELISA with: Human IL5Ralpha Human IL4R Rat IL9R Human IL13RA1 | |
Store at 4°C in the dark. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title