Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-15 Mouse anti-Human, Clone: 15G9, MilliporeSigma™
Mouse Monoclonal Antibody
$177.00 - $450.00
Specifications
Antigen | IL-15 |
---|---|
Clone | 15G9 |
Concentration | 1 mg/mL |
Applications | Immunohistochemistry (Paraffin), Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABF229825
|
MilliporeSigma
MABF229825UG |
25 μg |
Each for $177.00
|
|
|||||
MABF2298100
|
MilliporeSigma
MABF2298100UG |
100 μg |
Each for $450.00
|
|
|||||
Description
IL-15 Monoclonal antibody specifically detects IL-15 in Human samples. It is validated for Immunohistochemistry (Paraffin), Flow Cytometry.Specifications
IL-15 | |
1 mg/mL | |
Monoclonal | |
Purified | |
RUO | |
Purified mouse monoclonal antibody IgG1 in PBS with 0.05% sodium azide. | |
IgG1 κ | |
Protein G purified |
15G9 | |
Immunohistochemistry (Paraffin), Flow Cytometry | |
Unconjugated | |
Mouse | |
Human | |
Full-length mature recombinant human Interleukin-15 (without the signal peptide and propeptide). | |
Primary | |
Stable for 1 year at +2°C to +8°C from date of receipt. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title