Learn More
IL-15 Mouse anti-Human, DyLight 488, Clone: MM0368-13S34, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP211665G
Catalog No. NBP211665G
Add to cart
Description
Description
IL-15 Monoclonal antibody specifically detects IL-15 in Human samples. It is validated for Western Blot.
Specifications
Specifications
IL-15
MM0368-13S34
Western Blot
Purified
IL15
Produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with human recombinant protein of IL-15.
Protein G purified
Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research, Immune System Diseases, Immunology, Innate Immunity
Primary
3600
Western Blot
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
IL-15MGC9721, interleukin 15, interleukin-15
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human