Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-15R alpha Mouse anti-Human, PE/Cy5.5, Clone: JM7A4, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP143238PECY55
Description
IL-15R alpha Monoclonal antibody specifically detects IL-15R alpha in Human samples. It is validated for Flow Cytometry.Specifications
IL-15R alpha | |
JM7A4 | |
Flow Cytometry | |
Conjugated, Purified | |
IL15RA | |
The immunogen for this antibody was IL15Ra. | |
Protein A or G purified | |
Cancer | |
Primary | |
3601.0 |
Flow Cytometry | |
PE-Cyanine5.5 | |
PBS with 0.05% Sodium Azide | |
CD215 antigen, IL-15 receptor subunit alpha, IL-15RA, IL-15R-alpha, interleukin 15 receptor, alpha, interleukin-15 receptor subunit alpha, MGC104179 | |
Mouse | |
IgG2b | |
0.1 mL | |
Store at 4°C in the dark. Do not freeze. | |
Monoclonal | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title