Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-15R alpha Antibody (RM0103-6C38), mFluor Violet 450 SE, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP122466MFV450
Description
IL-15R alpha Monoclonal antibody specifically detects IL-15R alpha in Mouse samples. It is validated for Western BlotSpecifications
| IL-15R alpha | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot | |
| RM0103-6C38 | |
| Western Blot | |
| CD215 antigen, IL-15 receptor subunit alpha, IL-15RA, IL-15R-alpha, interleukin 15 receptor, alpha, interleukin-15 receptor subunit alpha, MGC104179 | |
| Mouse recombinant IL-15R alpha (extracellular domain) | |
| 0.1 mL | |
| Cancer | |
| 3601 | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction