Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-15R alpha Rat anti-Mouse, Clone: RM0103-6C38, Novus Biologicals™
Rat Monoclonal Antibody
$177.16 - $493.00
Specifications
Antigen | IL-15R alpha |
---|---|
Clone | RM0103-6C38 |
Concentration | LYOPH |
Dilution | Western Blot 1:500-1:1000 |
Applications | Western Blot |
Description
IL-15R alpha Monoclonal specifically detects IL-15R alpha in Mouse samples. It is validated for Western Blot.Specifications
IL-15R alpha | |
LYOPH | |
Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
3601 | |
IgG2 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
RM0103-6C38 | |
Western Blot 1:500-1:1000 | |
Monoclonal | |
Purified | |
RUO | |
CD215 antigen, IL-15 receptor subunit alpha, IL-15RA, IL-15R-alpha, interleukin 15 receptor, alpha, interleukin-15 receptor subunit alpha, MGC104179 | |
IL15RA | |
Primary | |
This was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant protein of the extracellular domain of IL-15RA. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title