Learn More
IL-15R alpha Mouse anti-Human, Janelia Fluor 549, Clone: JM7A4, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP143238JF549
Catalog No. NBP43238549
Add to cart
Description
Description
IL-15R alpha Monoclonal antibody specifically detects IL-15R alpha in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
IL-15R alpha
JM7A4
Flow Cytometry
Purified
IL15RA
The immunogen for this antibody was IL15Ra.
Protein A or G purified
Cancer
Primary
3601
Flow Cytometry
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
CD215 antigen, IL-15 receptor subunit alpha, IL-15RA, IL-15R-alpha, interleukin 15 receptor, alpha, interleukin-15 receptor subunit alpha, MGC104179
Mouse
IgG2b
0.1mL
Store at 4°C in the dark.
Monoclonal
Human