Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ IL-17/IL-17A Antibody Pair [HRP]

Mouse Monoclonal Antibody Pair

Manufacturer:  Novus Biologicals™ NBP2793845PLATES

 View more versions of this product

Catalog No. NB009808


Add to cart

Description

Description

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human IL-17/IL-17A.
Specifications

Specifications

HRP
Sandwich ELISA
1.17 pg/mL
3605
RUO
IL-17/IL-17A
Apoptosis, Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity
Human
IL17A
1. Capture Antibody: Dilute to a working concentration of 1 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.25 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 75 pg/mL is recommended.
Antibody Pairs
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human IL-17/IL-17A.
Sandwich ELISA
  • Standard: Each vial contains 4.7 ng of recombinant Human IL-17/IL-17A.
  • Capture Antibody: 1 mg/mL of mouse anti-Human IL-17/IL-17A monoclonal antibody (in PBS, pH 7.4).
  • Detection Antibody: 0.2 mg/mL of mouse anti-Human IL-17/IL-17A monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4C).
  • Storage is content dependent.
    Mouse
    5 Plates
    1.17-75 pg/mL
    CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, IL-17Acytotoxic T-lymphocyte-associated protein 8, IL-17CTLA-8, IL17interleukin-17A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8), interleukin 17A
    SDS
    Documents
    Product Certifications
    Promotions

    Promotions

    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit