Learn More
IL-17RE Rat anti-Mouse, Alexa Fluor 350, Clone: 944904, R&D Systems
Rat MonoclonalAntibody
Manufacturer:  R&D Systems FAB7997U100UG
Catalog No. FAB7997U100
Add to cart
This item is not returnable. View return policy
Description
Description
IL-17RE Monoclonal antibody specifically detects IL-17RE in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
IL-17RE
Monoclonal
Alexa Fluor 350
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
IL-17 RE, IL-17 receptor E, IL17RE, Il25r, interleukin 17 receptor E
Mouse myeloma cell line NSO-derived recombinant mouse IL-17 RE Met1-His414 Accession # Q8BH06
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects mouse IL-17 RE in direct ELISAs.
Flow Cytometry
944904
Flow Cytometry 0.25-1 ug/10^6 cells
Q8BH06
Rat
IgG1
RUO
Primary
132014
Mouse
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)