Learn More
IL-17RE Rat anti-Mouse, Alexa Fluor 647, Clone: 944904, R&D Systems
Rat Monoclonal Antibody
Manufacturer:  R&D Systems FAB7997R025UG
Catalog No. FAB7997R025
Add to cart
This item is not returnable. View return policy
Description
Description
IL-17RE Monoclonal antibody specifically detects IL-17RE in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
IL-17RE
944904
Flow Cytometry 0.1 ug/10^6 cells
Q8BH06
Rat
IgG1
25 ug
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
Detects mouse IL-17 RE in direct ELISAs.
Flow Cytometry
Alexa Fluor 647
Supplied in a saline solution containing BSA and Sodium Azide.
IL-17 RE, IL-17 receptor E, IL17RE, Il25r, interleukin 17 receptor E
Mouse myeloma cell line NS0-derived recombinant mouse IL-17 RE Met1-His414 Accession # Q8BH06
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
132014
Mouse