Learn More
IL-17 Receptor B Rat anti-Mouse, Alexa Fluor 647, Clone: 6B7, BD Pharmingen™
Rat Monoclonal Antibody
Manufacturer:  BD Biosciences 565866
Catalog No. BDB565866
Add to cart
Description
Description
IL-17 Receptor B Monoclonal antibody specifically detects IL-17 Receptor B in Mouse samples. It is validated for Flow cytometry.
Specifications
Specifications
IL-17 Receptor B
6B7
Alexa Fluor 647
Purified
Rat
Rat IgG1, κ
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to Alexa Fluor™ 647 under optimum conditions, and unreacted Alexa Fluor™ 647 was removed.
Monoclonal
Flow Cytometry
0.2mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
IL-17RB; IL-17B receptor; Il17rb; Evi27; Il17rb; IL-17ER; IL17RH1; IL-17Rh1
Mouse IL-17 Receptor B Recombinant Protein
50μg
Cell Biology
Primary
Murine
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Reseach Use Only.