Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-17B Mouse anti-Human, Janelia Fluor 646, Clone: MM0373-3D62, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP211672JF646
Description
IL-17B Monoclonal specifically detects IL-17B in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
IL-17B | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
IL17B | |
Human recombinant IL-17B | |
0.1 mL | |
Primary | |
Detects human IL-17B, but not IL-17A, C E or F. | |
Store at 4°C in the dark. | |
IgG2 |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
MM0373-3D62 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin | |
Cytokine Zcyto7, cytokine-like protein ZCYTO7, IL-17Binterleukin-17 beta, IL20, IL-20interleukin-17B, interleukin 17B, interleukin 20, Interleukin-20, MGC138900, MGC138901, neuronal interleukin-17 related factor, Neuronal interleukin-17-related factor, NIRF, ZCYTO7interleukin-20 | |
Mouse | |
Protein G purified | |
Biologically Active Proteins, Cytokine Research | |
27190 | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title