Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-17B Mouse anti-Human, PE-Cyanine7, Clone: MM0373-3D62, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP211672PECY7

Catalog No. N211672PEC7


Add to cart

Description

Description

IL-17B Monoclonal antibody specifically detects IL-17B in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

IL-17B
MM0373-3D62
Flow Cytometry
Conjugated, Purified
IL17B
Produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with human recombinant protein of IL-17B
Protein G purified
Biologically Active Proteins, Cytokine Research
Primary
27190.0
Flow Cytometry
PE-Cyanine7
PBS with 0.05% Sodium Azide
Cytokine Zcyto7, cytokine-like protein ZCYTO7, IL-17Binterleukin-17 beta, IL20, IL-20interleukin-17B, interleukin 17B, interleukin 20, Interleukin-20, MGC138900, MGC138901, neuronal interleukin-17 related factor, Neuronal interleukin-17-related factor, NIRF, ZCYTO7interleukin-20
Mouse
IgG1
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit