Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-17E/IL-25 Mouse anti-Human, Alexa Fluor™ 532, Clone: 8B1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP227324AF532

Catalog No. NP227324A32


Add to cart

Description

Description

IL-17E/IL-25 Monoclonal antibody specifically detects IL-17E/IL-25 in Human samples. It is validated for Western Blot, Flow Cytometry.
Specifications

Specifications

IL-17E/IL-25
8B1
Western Blot, Flow Cytometry
Conjugated, Purified
IL25
Full-length human IL17E recombinant protein was used as the immunogen for this antibody.
Protein G purified
Cytokine Research
Primary
64806.0
Flow Cytometry, Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
IL-17E, IL17Einterleukin-17E, IL-25, interleukin 17E, interleukin 25, Interleukin-17E, interleukin-25
Mouse
IgG1 Kappa
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit