Learn More
IL-17E/IL-25 Antibody (8B1), Alexa Fluor™ 594, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP227324AF594
Catalog No. NP227324A94
Add to Cart
Description
Description
IL-17E/IL-25 Monoclonal specifically detects IL-17E/IL-25 in Human samples. It is validated for Western Blot, Flow Cytometry.
Specifications
Specifications
IL-17E/IL-25
Monoclonal
Alexa Fluor 594
50 mM sodium borate with 0.05% sodium azide
IL25
Full-length human IL17E recombinant protein was used as the immunogen for this antibody.
Primary
Human
IgG1 κ
Western Blot, Flow Cytometry
8B1
Western Blot, Flow Cytometry
IL-17E, IL17Einterleukin-17E, IL-25, interleukin 17E, interleukin 25, Interleukin-17E, interleukin-25
Mouse
0.1 mL
64806
Store at 4°C in the dark.
Product Suggestions
Product Suggestions