Learn More
IL-17E/IL-25 Rat anti-Mouse, DyLight 550, Clone: RM0263-5J46, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP211677R
Catalog No. NBP211677R
Add to cart
Description
Description
IL-17E/IL-25 Monoclonal antibody specifically detects IL-17E/IL-25 in Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
IL-17E/IL-25
RM0263-5J46
Western Blot, Immunocytochemistry/Immunofluorescence
Purified
IL25
Produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant protein of IL-17E.
Protein G purified
Cytokine Research
Primary
64806
Immunocytochemistry, Immunofluorescence, Western Blot
DyLight 550
50mM Sodium Borate with 0.05% Sodium Azide
IL-17E, IL17Einterleukin-17E, IL-25, interleukin 17E, interleukin 25, Interleukin-17E, interleukin-25
Rat
IgG2
0.1 ml
Store at 4C in the dark.
Monoclonal
Mouse