Learn More
IL-17E/IL-25 Rat anti-Mouse, HRP, Clone: RM0263-5J46, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP211677H
Catalog No. NBP211677H
Add to cart
Description
Description
IL-17E/IL-25 Monoclonal antibody specifically detects IL-17E/IL-25 in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
IL-17E/IL-25
RM0263-5J46
Western Blot
Purified
IL25
Produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant protein of IL-17E.
Protein G purified
Cytokine Research
Primary
64806
Western Blot
HRP
PBS with No Preservative
IL-17E, IL17Einterleukin-17E, IL-25, interleukin 17E, interleukin 25, Interleukin-17E, interleukin-25
Rat
IgG2
0.1 ml
Store at 4C in the dark.
Monoclonal
Mouse