Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-17RA/IL-17R Mouse anti-Human, Alexa Fluor™ 594, Clone: 49M5D9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP225261AF594

Catalog No. NP22261A94


Add to cart

Description

Description

IL-17RA/IL-17R Monoclonal antibody specifically detects IL-17RA/IL-17R in Human samples. It is validated for Western Blot.
Specifications

Specifications

IL-17RA/IL-17R
49M5D9
Western Blot
Conjugated, Purified
IL17RA
Within the range of amino acids 186-264 of human IL-17RA were used as the immunogen for the antibody.
Protein G purified
Innate Immunity
Primary
23765.0
Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
CD217, CD217 antigen, CDw217interleukin 17 receptor, hIL-17R, IL-17 receptor A, IL-17RAMGC10262, IL17Rinterleukin-17 receptor A, interleukin 17 receptor A
Mouse
IgG2b Kappa
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit