Learn More
IL-17RA/IL-17R Mouse anti-Human, Alexa Fluor™ 750, Clone: 49M4D2, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP225258AF750
Catalog No. NP2228A70
Add to cart
Description
Description
IL-17RA/IL-17R Monoclonal antibody specifically detects IL-17RA/IL-17R in Human samples. It is validated for Western Blot,Immunohistochemistry,Immunohistochemistry (Paraffin).
Specifications
Specifications
IL-17RA/IL-17R
49M4D2
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
CD217, CD217 antigen, CDw217interleukin 17 receptor, hIL-17R, IL-17 receptor A, IL-17RAMGC10262, IL17Rinterleukin-17 receptor A, interleukin 17 receptor A
Mouse
IgG1 Kappa
Store at 4°C in the dark.
Monoclonal
Human
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
IL17RA
Within the range of amino acids 186-267 of human IL-17RA were used as the immunogen for the antibody.
0.1 mL
Primary
23765